accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOFFFOOH3C

CAS RN 100501-62-0

IUPAC Name: ethyl 1-ethyl-6,7,8-trifluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate
Synonyms: ethyl 1-ethyl-6,7,8-trifluoro-4-oxoquinoline-3-carboxylate
Molecular Weight (g/mol): 299.25
Molecular Formula: C14H12F3NO3
InChi Key: LWLLHOVWIFISMG-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN(CC)C2=C(F)C(F)=C(F)C=C2C1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results