accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 100516-88-9

IUPAC Name: (quinolin-6-yl)methanol
Synonyms: quinolin-6-ylmethanol
Molecular Weight (g/mol): 159.19
Molecular Formula: C10H9NO
InChi Key: YQEJIIUSNDZIGO-UHFFFAOYSA-N
SMILES: OCC1=CC=C2N=CC=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results