accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10052-47-8

IUPAC Name: triethyl(methyl)azanium chloride
Synonyms: triethyl(methyl)azanium chloride
Molecular Weight (g/mol): 151.68
Molecular Formula: C7H18ClN
InChi Key: NIUZJTWSUGSWJI-UHFFFAOYSA-M
SMILES: [Cl-].CC[N+](C)(CC)CC
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)