accessibility menu, dialog, popup

UserName

Viewing profile as user:
FeFeFeFeOPOOOPOOOOPOOOPOOOOPOOOPOOO

CAS RN 10058-44-3

IUPAC Name: tetrairon(3+) tri(phosphonatooxy)phosphonate
Synonyms: tetrairon(3+) tridiphosphate
Molecular Weight (g/mol): 745.2
Molecular Formula: Fe4O21P6
InChi Key: CADNYOZXMIKYPR-UHFFFAOYSA-B
SMILES: [Fe+3].[Fe+3].[Fe+3].[Fe+3].[O-]P([O-])(=O)OP([O-])([O-])=O.[O-]P([O-])(=O)OP([O-])([O-])=O.[O-]P([O-])(=O)OP([O-])([O-])=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (4)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (4)
  • (4)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (4)