accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOH3COOH3COOGd

CAS RN 100587-93-7

IUPAC Name: gadolinium(3+) triacetate
Synonyms: gadolinium(3+) triacetate
Molecular Weight (g/mol): 334.38
Molecular Formula: C6H9GdO6
InChi Key: LYQGMALGKYWNIU-UHFFFAOYSA-K
SMILES: [Gd+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (3)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)