accessibility menu, dialog, popup

UserName

Viewing profile as user:
FBrOH2N

CAS RN 1006-40-2

IUPAC Name: 2-bromo-4-fluorobenzamide
Synonyms: 2-bromo-4-fluorobenzamide
Molecular Weight (g/mol): 218.03
Molecular Formula: C7H5BrFNO
InChi Key: IWODMEGZVDWKPK-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(Br)C=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results