accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10060-13-6

IUPAC Name: diammonium tetrachlorocopperdiuide dihydrate
Synonyms: diammonium tetrachlorocuprate(2-) dihydrate
Molecular Weight (g/mol): 277.45
Molecular Formula: Cl4CuH12N2O2
InChi Key: LLMDTWKBBWCWNN-UHFFFAOYSA-L
SMILES: [NH4+].[NH4+].O.O.Cl[Cu--](Cl)(Cl)Cl
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)