accessibility menu, dialog, popup

UserName

Viewing profile as user:
GdOHOHOH

CAS RN 100634-91-1

IUPAC Name: gadolinium(3+) trihydroxide
Synonyms: gadolinium(3+) trihydroxide
Molecular Weight (g/mol): 208.27
Molecular Formula: GdH3O3
InChi Key: ILCLBMDYDXDUJO-UHFFFAOYSA-K
SMILES: [OH-].[OH-].[OH-].[Gd+3]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results