accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOONOOONOOOTm

CAS RN 100641-15-4

IUPAC Name: thulium(3+) trinitrate
Synonyms: thulium(3+) trinitrate
Molecular Weight (g/mol): 354.95
Molecular Formula: N3O9Tm
InChi Key: LLZBVBSJCNUKLL-UHFFFAOYSA-N
SMILES: [Tm+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)