CAS RN 100669-96-3
IUPAC Name:
sodium bis(fluorosulfonyl)azanide
Synonyms:
sodium bis(fluorosulfonyl)azanide
Molecular Weight (g/mol):
203.11
Molecular Formula:
F2NNaO4S2
InChi Key:
VCCATSJUUVERFU-UHFFFAOYSA-N
SMILES:
[Na+].FS(=O)(=O)[N-]S(F)(=O)=O
1
–
1
of
1
results
1
Sodium Bis(fluorosulfonyl)imide 98.0+%, TCI America™
CAS: 100669-96-3 Molecular Formula: F2NNaO4S2 Molecular Weight (g/mol): 203.11 InChI Key: VCCATSJUUVERFU-UHFFFAOYSA-N PubChem CID: 102310776 IUPAC Name: sodium;bis(fluorosulfonyl)azanide SMILES: [N-](S(=O)(=O)F)S(=O)(=O)F.[Na+]