accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1006876-28-3

IUPAC Name: N-ethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carboxamide
Synonyms: N-ethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carboxamide
Molecular Weight (g/mol): 276.14
Molecular Formula: C14H21BN2O3
InChi Key: TVMRHMZREIYXSR-UHFFFAOYSA-N
SMILES: CCNC(=O)C1=NC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results