accessibility menu, dialog, popup

UserName

Viewing profile as user:
HO

CAS RN 1007-03-0

IUPAC Name: cyclopropyl(phenyl)methanol
Synonyms: cyclopropyl(phenyl)methanol
Molecular Weight (g/mol): 148.21
Molecular Formula: C10H12O
InChi Key: GOXKCYOMDINCCD-UHFFFAOYNA-N
SMILES: OC(C1CC1)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results