accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1007-16-5

IUPAC Name: 3-bromo-4-fluorobenzoate
Synonyms: 3-bromo-4-fluorobenzoate
Molecular Weight (g/mol): 218
Molecular Formula: C7H3BrFO2
InChi Key: ONELILMJNOWXSA-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C(F)C(Br)=C1
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)