accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHNNH2NClN

CAS RN 1007-28-9

IUPAC Name: 6-chloro-N2-ethyl-1,3,5-triazine-2,4-diamine
Synonyms: deisopropylatrazine 6-deisopropylatrazine caswell no. 033F C5H8ClN5 desisopropylatrazine
Molecular Weight (g/mol): 173.6
Molecular Formula: C5H8ClN5
InChi Key: IVENSCMCQBJAKW-UHFFFAOYSA-N
SMILES: CCNC1=NC(N)=NC(Cl)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results