accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO

CAS RN 1007-32-5

IUPAC Name: 1-phenylbutan-2-one
Synonyms: 1-phenyl-2-butanone
Molecular Weight (g/mol): 148.21
Molecular Formula: C10H12O
InChi Key: GKDLTXYXODKDEA-UHFFFAOYSA-N
SMILES: CCC(=O)CC1=CC=CC=C1
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)