accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10072-05-6

IUPAC Name: methyl 2-(dimethylamino)benzoate
Synonyms: methyl 2-(dimethylamino)benzoate
Molecular Weight (g/mol): 179.22
Molecular Formula: C10H13NO2
InChi Key: ZCNSBHAIPOWHJE-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC=C1N(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results