accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10075-51-1

BrN

CAS RN 10075-51-1

IUPAC Name: 1-benzyl-5-bromo-1H-indole
Synonyms: 1-benzyl-5-bromoindole
Molecular Weight (g/mol): 286.17
Molecular Formula: C15H12BrN
InChi Key: AQXJFUYUNHLBGU-UHFFFAOYSA-N
SMILES: BrC1=CC=C2N(CC3=CC=CC=C3)C=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results