accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10076-48-9

CH3OOCH3OH3COH3C

CAS RN 10076-48-9

IUPAC Name: methyl 2,2-dimethoxypropanoate
Synonyms: methyl 2,2-dimethoxypropanoate
Molecular Weight (g/mol): 148.16
Molecular Formula: C6H12O4
InChi Key: KHQQDBIXHUJARJ-UHFFFAOYSA-N
SMILES: COC(=O)C(C)(OC)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results