accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CBrOH

CAS RN 100760-04-1

IUPAC Name: 1-(4-bromophenyl)ethan-1-ol
Synonyms: 1-(4-bromophenyl)ethanol
Molecular Weight (g/mol): 201.06
Molecular Formula: C8H9BrO
InChi Key: XTDTYSBVMBQIBT-UHFFFAOYNA-N
SMILES: CC(O)C1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results