accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1007847-76-8

IUPAC Name: 4,4,5,5-tetramethyl-2-{4-[(thiophen-2-yl)methyl]phenyl}-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-[4-(thiophen-2-ylmethyl)phenyl]-1,3,2-dioxaborolane
Molecular Weight (g/mol): 300.22
Molecular Formula: C17H21BO2S
InChi Key: OPJUSDABOPHYPQ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(CC2=CC=CS2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results