accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1008-76-0

IUPAC Name: 5-phenyloxolan-2-one
Synonyms: 5-phenyloxolan-2-one
Molecular Weight (g/mol): 162.19
Molecular Formula: C10H10O2
InChi Key: AEUULUMEYIPECD-UHFFFAOYNA-N
SMILES: O=C1CCC(O1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results