accessibility menu, dialog, popup

UserName

Viewing profile as user:
N

CAS RN 1008-89-5

IUPAC Name: 2-phenylpyridine
Synonyms: 2-phenylpyridine
Molecular Weight (g/mol): 155.2
Molecular Formula: C11H9N
InChi Key: VQGHOUODWALEFC-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=CC=CC=N1
Boiling Point 
  • (4)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (2)
  • (2)
  • (4)
Quantity 
  • (2)
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (6)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)