accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10081-67-1

IUPAC Name: 4-(2-phenylpropan-2-yl)-N-[4-(2-phenylpropan-2-yl)phenyl]aniline
Synonyms: 4-(2-phenylpropan-2-yl)-N-[4-(2-phenylpropan-2-yl)phenyl]aniline
Molecular Weight (g/mol): 405.59
Molecular Formula: C30H31N
InChi Key: UJAWGGOCYUPCPS-UHFFFAOYSA-N
SMILES: CC(C)(C1=CC=CC=C1)C1=CC=C(NC2=CC=C(C=C2)C(C)(C)C2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results