accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 100836-85-9

IUPAC Name: 2-(benzyloxy)propanoic acid
Synonyms: 2-(benzyloxy)propanoic acid
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: XWAVPOFYNPXXEL-UHFFFAOYNA-N
SMILES: CC(OCC1=CC=CC=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results