accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNFFF

CAS RN 100846-24-0

IUPAC Name: 5-(trifluoromethyl)-1H-indole
Synonyms: 5-(trifluoromethyl)-1H-indole
Molecular Weight (g/mol): 185.15
Molecular Formula: C9H6F3N
InChi Key: LCFDJWUYKUPBJM-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C2NC=CC2=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)