accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1009-93-4

CH3SiH3CNHSiCH3CH3NHSiH3CH3CHN

CAS RN 1009-93-4

IUPAC Name: 2,2,4,4,6,6-hexamethyl-1,3,5,2,4,6-triazatrisilinane
Synonyms: 2,2,4,4,6,6-hexamethyl-1,3,5,2,4,6-triazatrisilinane
Molecular Weight (g/mol): 219.51
Molecular Formula: C6H21N3Si3
InChi Key: WGGNJZRNHUJNEM-UHFFFAOYSA-N
SMILES: C[Si]1(C)N[Si](C)(C)N[Si](C)(C)N1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)