accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1009303-56-3

IUPAC Name: [4-fluoro-3-(2H-1,2,3,4-tetrazol-5-yl)phenyl]boronic acid
Synonyms: 4-fluoro-3-(2H-1,2,3,4-tetrazol-5-yl)phenylboronic acid
Molecular Weight (g/mol): 207.96
Molecular Formula: C7H6BFN4O2
InChi Key: FELFJHZMQYCKPT-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(C2=NNN=N2)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results