accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10094-58-3

OH(R)O(S)O(R)O(S)O(R)O(R)O(S)OHO(R)OH(S)OH(S)OH(R)OH(S)HO(S)HO(R)HO(S)HO(R)HO(R)HO(S)HO(R)OH

CAS RN 10094-58-3

IUPAC Name: (2S,3R,4S,5R,6R)-2-{[(2R,3R,4S,5R,6S)-6-{[(2R,3S,4S,5R,6R)-6-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-3,4,5-trihydroxyoxan-2-yl]methoxy}-3,4,5-trihydroxyoxan-2-yl]methoxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms: (2S,3R,4S,5R,6R)-2-{[(2R,3R,4S,5R,6S)-6-{[(2R,3S,4S,5R,6R)-6-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-3,4,5-trihydroxyoxan-2-yl]methoxy}-3,4,5-trihydroxyoxan-2-yl]methoxy}-6-(hydroxymethyl)oxane-3,4,5-triol stachyose stachyose tetrahydrate stachyose hydrate D-stachyose stachyose (substance)
Molecular Weight (g/mol): 666.58
Molecular Formula: C24H42O21
InChi Key: UQZIYBXSHAGNOE-XNSRJBNMSA-N
SMILES: OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO[C@H]3O[C@H](CO[C@H]4O[C@H](CO)[C@H](O)[C@H](O)[C@H]4O)[C@H](O)[C@H](O)[C@H]3O)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (1)
  • (1)