accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH2HCl

CAS RN 100959-19-1

IUPAC Name: 4-methylcyclohexan-1-amine hydrochloride
Synonyms: 4-methylcyclohexan-1-amine hydrochloride
Molecular Weight (g/mol): 149.66
Molecular Formula: C7H16ClN
InChi Key: GIRKJSRZELQHDX-UHFFFAOYSA-N
SMILES: Cl.CC1CCC(N)CC1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results