accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10099-67-9

IUPAC Name: lutetium(3+) trinitrate
Synonyms: lutetium(3+) trinitrate
Molecular Weight (g/mol): 360.98
Molecular Formula: LuN3O9
InChi Key: APRNQTOXCXOSHO-UHFFFAOYSA-N
SMILES: [Lu+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Grade 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results