accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CN(R)(R)HN

CAS RN 100999-26-6

IUPAC Name: (2R,11R)-10-methyl-10,15-diazapentacyclo[11.6.1.0²,¹¹.0³,⁸.0¹⁶,²⁰]icosa-1(20),3,5,7,13,16,18-heptaene
Synonyms: (2R,11R)-10-methyl-10,15-diazapentacyclo[11.6.1.0²,¹¹.0³,⁸.0¹⁶,²⁰]icosa-1(20),3,5,7,13,16,18-heptaene
Molecular Weight (g/mol): 274.37
Molecular Formula: C19H18N2
InChi Key: WRNKIDLXXXIELU-IEBWSBKVSA-N
SMILES: CN1CC2=CC=CC=C2[C@H]2[C@H]1CC1=CNC3=CC=CC2=C13

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results