accessibility menu, dialog, popup

UserName

Viewing profile as user:
OPOO

CAS RN 101-02-0

IUPAC Name: triphenyl phosphite
Synonyms: triphenyl phosphite
Molecular Weight (g/mol): 310.29
Molecular Formula: C18H15O3P
InChi Key: HVLLSGMXQDNUAL-UHFFFAOYSA-N
SMILES: O(P(OC1=CC=CC=C1)OC1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (6)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (1)
  • (5)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (4)