accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNClNNHClN

CAS RN 101-05-3

IUPAC Name: 4,6-dichloro-N-(2-chlorophenyl)-1,3,5-triazin-2-amine
Synonyms: anilazine
Molecular Weight (g/mol): 275.52
Molecular Formula: C9H5Cl3N4
InChi Key: IMHBYKMAHXWHRP-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1NC1=NC(Cl)=NC(Cl)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results