accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 101-18-8

IUPAC Name: 3-(phenylamino)phenol
Synonyms: phenol, 3-(phenylamino)-
Molecular Weight (g/mol): 185.23
Molecular Formula: C12H11NO
InChi Key: NDACNGSDAFKTGE-UHFFFAOYSA-N
SMILES: OC1=CC=CC(NC2=CC=CC=C2)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results