CAS RN 101-27-9
IUPAC Name:
4-chlorobut-2-yn-1-yl N-(3-chlorophenyl)carbamate
Synonyms:
barban
Molecular Weight (g/mol):
258.1
Molecular Formula:
C11H9Cl2NO2
InChi Key:
MCOQHIWZJUDQIC-UHFFFAOYSA-N
SMILES:
ClCC#CCOC(=O)NC1=CC=CC(Cl)=C1
1
–
1
of
1
results
1
Barban, SPEX CertiPrep™
CAS: 101-27-9 Molecular Formula: C11H9Cl2NO2 Molecular Weight (g/mol): 258.098 InChI Key: MCOQHIWZJUDQIC-UHFFFAOYSA-N PubChem CID: 7551 ChEBI: CHEBI:33421 IUPAC Name: 4-chlorobut-2-ynyl N-(3-chlorophenyl)carbamate SMILES: C1=CC(=CC(=C1)Cl)NC(=O)OCC#CCCl