accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 101-54-2

IUPAC Name: N1-phenylbenzene-1,4-diamine
Synonyms: N-phenyl-p-phenylenediamine azosalt R p-semidine N-phenyl-P-phenylenediamine p-aminodiphenylamine N-4'-bianiline 4-aminodiphenylamine luxan black R
Molecular Weight (g/mol): 184.24
Molecular Formula: C12H12N2
InChi Key: ATGUVEKSASEFFO-UHFFFAOYSA-N
SMILES: NC1=CC=C(NC2=CC=CC=C2)C=C1
Molecular Weight (g/mol) 
  • (5)
  • (4)
Percent Purity 
  • (2)
  • (7)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (4)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (5)
  • (4)
  • (2)
  • (7)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (3)
  • (2)
  • (3)