accessibility menu, dialog, popup

UserName

Viewing profile as user:
O

CAS RN 101-84-8

IUPAC Name: phenoxybenzene
Synonyms: diphenyl-ether phenyl oxide geranium crystals biphenyl oxide diphenylaether diphenyloxid diphenyl oxide chemcryl JK-EB
Molecular Weight (g/mol): 170.21
Molecular Formula: C12H10O
InChi Key: USIUVYZYUHIAEV-UHFFFAOYSA-N
SMILES: O(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (8)
  • (2)
  • (6)
Boiling Point 
  • (2)
  • (3)
  • (3)
  • (2)
  • (5)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (8)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (1)
  • (4)
  • (1)
  • (2)
  • (8)
  • (8)
  • (2)
  • (6)
  • (2)
  • (3)
  • (3)
  • (2)
  • (5)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (8)
  • (2)
  • (1)
  • (1)
  • (1)