CAS RN 101-91-7
IUPAC Name:
N-(4-hydroxyphenyl)butanamide
Synonyms:
N-(4-hydroxyphenyl)butanamide
Molecular Weight (g/mol):
179.22
Molecular Formula:
C10H13NO2
InChi Key:
KESXDDATSRRGAH-UHFFFAOYSA-N
SMILES:
CCCC(=O)NC1=CC=C(O)C=C1
1
–
1
of
1
results
1
Sigma Aldrich 4'-Hydroxybutyranilide
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.