CAS RN 1010-95-3
5-Methyltryptamine, hydrochloride, 98%, Thermo Scientific™
Molecular Formula: C11H15N2 Molecular Weight (g/mol): 175.25 MDL Number: MFCD00012683 InChI Key: PYOUAIQXJALPKW-UHFFFAOYSA-O Synonym: 5-methyltryptamine hydrochloride,2-5-methyl-1h-indol-3-yl ethanamine hydrochloride,3-2-aminoethyl-5-methylindole hydrochloride,5-methyl-1h-indole-3-ethylamine monohydrochloride,5-methyltryptamine hcl,1h-indole-3-ethanamine, 5-methyl-, monohydrochloride,2-5-methylindol-3-yl ethylamine, chloride,5-methyltryptaminehydrochloride,5-methyl-1h-indole-3-ethylamine hcl,pubchem24287 PubChem CID: 6917145 SMILES: CC1=CC=C2NC=C(CC[NH3+])C2=C1