accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10101-58-3

IUPAC Name: λ²-cobalt(2+) tungsten tetraoxidandiide
Synonyms: λ²-cobalt(2+) tungsten tetraoxidandiide
Molecular Weight (g/mol): 306.77
Molecular Formula: CoO4W
InChi Key: BGLJOHZOBUGBBY-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[O--].[Co++].[W]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results