CAS RN 10101-63-0
Lead(II) iodide, 99.9985% (metals basis)
CAS: 10101-63-0 Molecular Formula: I2Pb Molecular Weight (g/mol): 461.00 MDL Number: MFCD00011163 InChI Key: RQQRAHKHDFPBMC-UHFFFAOYSA-L Synonym: lead ii iodide,lead diiodide,plumbous iodide,lead iodide pbi2,pbi2,plumbum jodatum,blei ii-iodid,hsdb 636,diiodo-,e2-plumbane PubChem CID: 24931 IUPAC Name: λ²-lead(2+) diiodide SMILES: [I-].[I-].[Pb++]
Lead(II) iodide, ultra dry, 99.999% (metals basis)
CAS: 10101-63-0 Molecular Formula: I2Pb Molecular Weight (g/mol): 461.00 MDL Number: MFCD00011163 InChI Key: RQQRAHKHDFPBMC-UHFFFAOYSA-L Synonym: lead ii iodide,lead diiodide,plumbous iodide,lead iodide pbi2,pbi2,plumbum jodatum,blei ii-iodid,hsdb 636,diiodo-,e2-plumbane PubChem CID: 24931 IUPAC Name: λ²-lead(2+) diiodide SMILES: [I-].[I-].[Pb++]
Lead(II) iodide, 99%
CAS: 10101-63-0 Molecular Formula: I2Pb Molecular Weight (g/mol): 461.00 MDL Number: MFCD00011163 InChI Key: RQQRAHKHDFPBMC-UHFFFAOYSA-L Synonym: lead ii iodide,lead diiodide,plumbous iodide,lead iodide pbi2,pbi2,plumbum jodatum,blei ii-iodid,hsdb 636,diiodo-,e2-plumbane PubChem CID: 24931 IUPAC Name: λ²-lead(2+) diiodide SMILES: [I-].[I-].[Pb++]
Lead(II) iodide, 98.5%
CAS: 10101-63-0 Molecular Formula: I2Pb Molecular Weight (g/mol): 461.00 MDL Number: MFCD00011163 InChI Key: RQQRAHKHDFPBMC-UHFFFAOYSA-L Synonym: lead ii iodide,lead diiodide,plumbous iodide,lead iodide pbi2,pbi2,plumbum jodatum,blei ii-iodid,hsdb 636,diiodo-,e2-plumbane PubChem CID: 24931 IUPAC Name: λ²-lead(2+) diiodide SMILES: [I-].[I-].[Pb++]
Lead(II) Iodide (99.99%, trace metals basis) 98.0+%, TCI America™
CAS: 10101-63-0 Molecular Formula: I2Pb Molecular Weight (g/mol): 461.00 MDL Number: MFCD00011163 InChI Key: RQQRAHKHDFPBMC-UHFFFAOYSA-L Synonym: lead ii iodide,lead diiodide,plumbous iodide,lead iodide pbi2,pbi2,plumbum jodatum,blei ii-iodid,hsdb 636,diiodo-,e2-plumbane PubChem CID: 24931 IUPAC Name: λ²-lead(2+) diiodide SMILES: [I-].[I-].[Pb++]
Sigma Aldrich Lead(II) iodide
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Lead(II) iodide
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Lead(II) iodide
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Lead(II) iodide
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.