accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10101-98-1

IUPAC Name: nickel(2+) heptahydrate sulfate
Synonyms: nickel sulfate heptahydrate nickelous sulfate heptahydrate nickel(II) sulfate heptahydrate
Molecular Weight (g/mol): 280.85
Molecular Formula: H14NiO11S
InChi Key: OGKAGKFVPCOHQW-UHFFFAOYSA-L
SMILES: O.O.O.O.O.O.O.[Ni++].[O-]S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)