accessibility menu, dialog, popup

UserName

Viewing profile as user:
SNClOHO

CAS RN 101012-12-8

IUPAC Name: 2-chloro-1,3-thiazole-5-carboxylic acid
Synonyms: 2-chloro-1,3-thiazole-5-carboxylic acid
Molecular Weight (g/mol): 163.58
Molecular Formula: C4H2ClNO2S
InChi Key: HNJOKQPEJIWTRF-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=C(Cl)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results