CAS RN 10102-45-1
Thallium(I) nitrate, 99.99%, (trace metal basis)
CAS: 10102-45-1 Molecular Formula: NO3Tl Molecular Weight (g/mol): 266.38 MDL Number: MFCD00011280 InChI Key: FYWSTUCDSVYLPV-UHFFFAOYSA-N Synonym: thallium i nitrate,thallium 1+ nitrate,thallous nitrate,thallium mononitrate,thallium nitrate van,thallium nitrate, tlno3,unii-ljq38dsr12,rcra waste number u217,thallium i nitrate 1:1 PubChem CID: 24937 IUPAC Name: thallium(1+);nitrate SMILES: [Tl+].[O-][N+]([O-])=O
Thallium(I) nitrate, 99.5%
CAS: 10102-45-1 Molecular Formula: NO3Tl Molecular Weight (g/mol): 266.38 MDL Number: MFCD00011280 InChI Key: FYWSTUCDSVYLPV-UHFFFAOYSA-N Synonym: thallium i nitrate,thallium 1+ nitrate,thallous nitrate,thallium mononitrate,thallium nitrate van,thallium nitrate, tlno3,unii-ljq38dsr12,rcra waste number u217,thallium i nitrate 1:1 PubChem CID: 24937 IUPAC Name: thallium(1+);nitrate SMILES: [Tl+].[O-][N+]([O-])=O
Thallium Standard For ICP, 1000μg/mL in 2% HNO3, SPEX CertiPrep™
CAS: 10102-45-1 Molecular Formula: NO3Tl Molecular Weight (g/mol): 266.38 MDL Number: MFCD00011280 InChI Key: FYWSTUCDSVYLPV-UHFFFAOYSA-N PubChem CID: 24937 IUPAC Name: thallium(1+);nitrate SMILES: [Tl+].[O-][N+]([O-])=O
VeriSpec™ Thallium Standard for ICP/MS 100 ppm in 2% HNO3, Ricca Chemical
CAS: 10102-45-1 Molecular Formula: NO3Tl Molecular Weight (g/mol): 266.38 MDL Number: MFCD00011280 InChI Key: FYWSTUCDSVYLPV-UHFFFAOYSA-N PubChem CID: 24937 IUPAC Name: λ1-thallanylium nitrate SMILES: [Tl+].[O-][N+]([O-])=O
VeriSpec™ Thallium Standard for ICP 1000 ppm in 2% HNO3, Ricca Chemical
CAS: 10102-45-1 Molecular Formula: NO3Tl Molecular Weight (g/mol): 266.38 MDL Number: MFCD00011280 InChI Key: FYWSTUCDSVYLPV-UHFFFAOYSA-N PubChem CID: 24937 IUPAC Name: λ1-thallanylium nitrate SMILES: [Tl+].[O-][N+]([O-])=O
VeriSpec™ Thallium (Tl) Standard for AAS 1000 ppm in 2% HNO3, Ricca Chemical
CAS: 10102-45-1 Molecular Formula: NO3Tl Molecular Weight (g/mol): 266.38 MDL Number: MFCD00011280 InChI Key: FYWSTUCDSVYLPV-UHFFFAOYSA-N PubChem CID: 24937 IUPAC Name: λ1-thallanylium nitrate SMILES: [Tl+].[O-][N+]([O-])=O
VeriSpec™ Thallium Standard for ICP/MS 10 ppm in 2% HNO3, Ricca Chemical
CAS: 10102-45-1 Molecular Formula: NO3Tl Molecular Weight (g/mol): 266.38 MDL Number: MFCD00011280 InChI Key: FYWSTUCDSVYLPV-UHFFFAOYSA-N PubChem CID: 24937 IUPAC Name: λ1-thallanylium nitrate SMILES: [Tl+].[O-][N+]([O-])=O
Thallium Standard For ICP/MS, SPEX CertiPrep™
CAS: 10102-45-1 Molecular Formula: NO3Tl Molecular Weight (g/mol): 266.38 MDL Number: MFCD00011280 InChI Key: FYWSTUCDSVYLPV-UHFFFAOYSA-N PubChem CID: 24937 IUPAC Name: thallium(1+);nitrate SMILES: [Tl+].[O-][N+]([O-])=O
Sigma Aldrich Thallium(I) nitrate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Thallium(I) nitrate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Ricca Chemical Company Thallium Standard, 1mL= 1mg Tl, 1000ppm Tl, Ricca Chemical
CAS: 10102-45-1 Molecular Formula: NO3Tl Molecular Weight (g/mol): 266.38 MDL Number: MFCD00011280 InChI Key: FYWSTUCDSVYLPV-UHFFFAOYSA-N PubChem CID: 24937 IUPAC Name: λ¹-thallanylium nitrate SMILES: [Tl+].[O-][N+]([O-])=O