accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10108-86-8

ClH3CNCH3CH3H3C

CAS RN 10108-86-8

IUPAC Name: trimethyl(octyl)azanium chloride
Synonyms: octyltrimethylammonium chloride
Molecular Weight (g/mol): 207.79
Molecular Formula: C11H26ClN
InChi Key: AQZSPJRLCJSOED-UHFFFAOYSA-M
SMILES: [Cl-].CCCCCCCC[N+](C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)