accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 101093-56-5

IUPAC Name: 4-(benzyloxy)-2-methylbenzaldehyde
Synonyms: 4-(benzyloxy)-2-methylbenzaldehyde
Molecular Weight (g/mol): 226.28
Molecular Formula: C15H14O2
InChi Key: OGVPJBJWJCZBTH-UHFFFAOYSA-N
SMILES: CC1=C(C=O)C=CC(OCC2=CC=CC=C2)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results