accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1011-17-2

IUPAC Name: 2-(piperazin-1-yl)phenol
Synonyms: 2-(piperazin-1-yl)phenol
Molecular Weight (g/mol): 178.24
Molecular Formula: C10H14N2O
InChi Key: UORNTHBBLYBAJJ-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1N1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results