accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOHN

CAS RN 1011-65-0

IUPAC Name: methyl 1H-indole-5-carboxylate
Synonyms: INDOLE5carboxylicacidmethylester
Molecular Weight (g/mol): 175.19
Molecular Formula: C10H9NO2
InChi Key: DRYBMFJLYYEOBZ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C2NC=CC2=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)