accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10111-08-7

IUPAC Name: 1H-imidazole-2-carbaldehyde
Synonyms: 1H-imidazole-2-carbaldehyde
Molecular Weight (g/mol): 96.09
Molecular Formula: C4H4N2O
InChi Key: XYHKNCXZYYTLRG-UHFFFAOYSA-N
SMILES: O=CC1=NC=CN1
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (3)
  • (2)