accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHNOO

CAS RN 10112-15-9

IUPAC Name: N-ethyl-2-nitroaniline
Synonyms: N-ethyl-2-nitroaniline
Molecular Weight (g/mol): 166.18
Molecular Formula: C8H10N2O2
InChi Key: CQIKVOWCSGXCCG-UHFFFAOYSA-N
SMILES: CCNC1=CC=CC=C1[N+]([O-])=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results